Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagents

Catalog No. 01335432
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-432 1 L
01-335-431 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-432 Supplier Reagents Holdings Llc Supplier No. CD1001001A
 Restricted Item
Add to Cart
Add to Cart

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Doctor Solution (Sodium Plumbite) ASTM D 4952
Color Colorless with Slight Orange
CAS 7732-18-5, 1310-73-2
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.14
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.