Learn More
Medchemexpress LLC Docusate Sodium (Dioctyl sulfosuccinate sodium salt) | 577-11-7 | MFCD00012455 | 98.0% | 500 MG
SDP
Catalog No. 5000411848
Encompass_Preferred
Docusate Sodium is a sulfated surfactant that is a main component in stool softeners. It may inactivate viral pathogens by disrupting viral envelopes and/or denaturing/disassociating proteins. It has shown effectiveness in vitro against wild type and drug-resistant strains of HSV type 1 and 2. Docusate Sodium is also identified as an obesogen; developmental exposure to it in mice leads to increased adult adiposity, inflammation, metabolic disorder, and dyslipidemia in offspring fed a standard diet.
  • Main component in stool softeners
  • Inactivates viral pathogens by disrupting viral envelopes and/or denaturing proteins
  • Effective against wild type and drug-resistant strains of HSV type 1 and 2
  • Identified as an obesogen
Catalog No. 50-004-11848 Supplier Medchemexpress LLC Supplier No. HYB1268500MG
Docusate Sodium is a sulfated surfactant that is a main component in stool softeners. It may inactivate viral pathogens by disrupting viral envelopes and/or denaturing/disassociating proteins. It has shown effectiveness in vitro against wild type and drug-resistant strains of HSV type 1 and 2. Docusate Sodium is also identified as an obesogen; developmental exposure to it in mice leads to increased adult adiposity, inflammation, metabolic disorder, and dyslipidemia in offspring fed a standard diet.
  • Main component in stool softeners
  • Inactivates viral pathogens by disrupting viral envelopes and/or denaturing proteins
  • Effective against wild type and drug-resistant strains of HSV type 1 and 2
  • Identified as an obesogen