Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Docusate sodium salt

Catalog No. 86140500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-140-500G 500 g
86-140-100G 100 g
86-140-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 86-140-500G Supplier Sigma Aldrich Supplier No. 86140500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Docusate sodium salt is an emollient which can combine with the active pharmaceutical ingredients (APIs) such as ranitidine hydrochloride and lidocaine hydrochloride to form hydrophobic ionic liquids (IL-APIs)).Forms reverse micelles in hydrocarbon solvents; Suitable for the solubilization of the major myelin transmembrane proteolipid

Specifications

CAS 577-11-7
Quantity 500 g
Molecular Formula C20H37NaO7S
Linear Formula C20H37NaO7S
MDL Number MFCD00012455
Synonym AOT; Bis(2-ethylhexyl) sulfosuccinate sodium salt; Sodium bis(2-ethylhexyl) sulfosuccinate; Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt; Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt; Dioctyl sulfosuccinate sodium salt
Molecular Weight (g/mol) 444.56
Percent Purity ≥96.0% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.