Learn More
Medchemexpress LLC DODAP | 127512-29-2 | 99.7% | 100 MG
SDP
Catalog No. 5000340844
Encompass_Preferred
DODAP is a cationic lipid commonly used as a lipid component in liposomes. It is designed for encapsulating siRNA and delivering immunostimulated chemotherapeutic agents, effective both in vitro and in vivo.
  • Cationic lipid component in liposomes
  • Used for encapsulating siRNA
  • Delivers immunostimulated chemotherapeutic agents
  • Applicable in vitro and in vivo
  • Shows promise for research in vaccines and inflammation
Catalog No. 50-003-40844 Supplier Medchemexpress LLC Supplier No. HY130751100MG
DODAP is a cationic lipid commonly used as a lipid component in liposomes. It is designed for encapsulating siRNA and delivering immunostimulated chemotherapeutic agents, effective both in vitro and in vivo.
  • Cationic lipid component in liposomes
  • Used for encapsulating siRNA
  • Delivers immunostimulated chemotherapeutic agents
  • Applicable in vitro and in vivo
  • Shows promise for research in vaccines and inflammation