Learn More
Medchemexpress LLC Dodap | 127512-29-2 | MFCD01321074 | 98.0% | 648.05 g/mol | C41H77NO4 | 5 MG
SDP
Catalog No. 5000262301
Encompass_Preferred
DODAP is an ionizable cationic lipid used as the lipid component in liposomal formulations for nucleic acid delivery and related research. It is commonly employed to encapsulate siRNA and support intracellular delivery of therapeutic or immunostimulatory agents in vitro and in vivo, with applications in vaccine and inflammation studies.
  • Cationic lipid that facilitates siRNA encapsulation and delivery.
  • Ionizable with a pKa around 5.59 to aid endosomal release.
  • High purity (98.0%) supplied as a liquid for formulation flexibility.
  • Available in multiple pack sizes suitable for small-scale experiments.
  • Defined storage conditions for long-term stability in pure form and in solvent.
Catalog No. 50-002-62301 Supplier Medchemexpress LLC Supplier No. HY1307515MG
DODAP is an ionizable cationic lipid used as the lipid component in liposomal formulations for nucleic acid delivery and related research. It is commonly employed to encapsulate siRNA and support intracellular delivery of therapeutic or immunostimulatory agents in vitro and in vivo, with applications in vaccine and inflammation studies.
  • Cationic lipid that facilitates siRNA encapsulation and delivery.
  • Ionizable with a pKa around 5.59 to aid endosomal release.
  • High purity (98.0%) supplied as a liquid for formulation flexibility.
  • Available in multiple pack sizes suitable for small-scale experiments.
  • Defined storage conditions for long-term stability in pure form and in solvent.