Learn More
Medchemexpress LLC HY-130751 50mg , DODAP CAS:127512-29-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13075150MG
Encompass_Preferred
HY-130751 50mg DODAP CAS: 127512-29-2 DODAP is a cationic lipid. The ionizable lipid DODAP is a lipid component of the liposome. DODAP can be used to encapsulate siRNA, immunostimulatory chemotherapeutic agents for in vitro and in vivo delivery and so on. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1356