Learn More
Medchemexpress LLC Dodeca-2E,4E,8Z,10Z/E-N-tetraenoic acid isobutylamide | 866602-52-0 | 98.5% | 247.38 | C16H25NO | 5mg
SDP
Catalog No. 5000270158
Encompass_Preferred
Dodeca-2E 4E 8Z 10Z/E-N-tetraenoic acid isobutylamide is a brain-penetrant substance and the main alkamide in Echinacea preparations which might be used for research of common cold and various upper respiratory infections[1]
Catalog No. 50-002-70158 Supplier Medchemexpress LLC Supplier No. HYW7098565MG
Dodeca-2E 4E 8Z 10Z/E-N-tetraenoic acid isobutylamide is a brain-penetrant substance and the main alkamide in Echinacea preparations which might be used for research of common cold and various upper respiratory infections[1]