Learn More
Medchemexpress LLC Dodecaethylene glycol | 6790-09-6 | 99.9% | C24H50O13 | 250 MG
SDP
Catalog No. 5000352781
Encompass_Preferred
Dodecaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules that comprise two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. These compounds function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • Used in the synthesis of PROTACs
  • Functions by leveraging the intracellular ubiquitin-proteasome system
  • Selectively degrades specific target proteins
  • For research use only
Catalog No. 50-003-52781 Supplier Medchemexpress LLC Supplier No. 250MGHY141233
Add to Cart
Add to Cart
Dodecaethylene glycol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules that comprise two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. These compounds function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • Used in the synthesis of PROTACs
  • Functions by leveraging the intracellular ubiquitin-proteasome system
  • Selectively degrades specific target proteins
  • For research use only