Learn More
Medchemexpress LLC Dodecaethylene glycol | 6790-09-6 | 99.93% | C24H50O13 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1412331G
Encompass_Preferred
Dodecaethylene glycol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to contain two different ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • High purity (99.93%) verified by HPLC
  • For research use only
  • Suitable for laboratory chemical applications
  • Appears as a white to off-white solid
Catalog No. 50-003-53831