Learn More
Medchemexpress LLC Dodecaethylene glycol | 6790-09-6 | MFCD06201001 | 99.9% | C24H50O13 | 100 G
SDP
Catalog No. 5000354248
Encompass_Preferred
Dodecaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker.
  • Useful in the synthesis of PROTACs.
  • Enables targeted protein degradation via the ubiquitin-proteasome system.
  • High purity of 99.9%.
  • Appears as a white to off-white solid.
Catalog No. 50-003-54248 Supplier Medchemexpress LLC Supplier No. HY141233100G
Dodecaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker.
  • Useful in the synthesis of PROTACs.
  • Enables targeted protein degradation via the ubiquitin-proteasome system.
  • High purity of 99.9%.
  • Appears as a white to off-white solid.