Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecaethylene Glycol 90.0+%, TCI America™
SDP

Catalog No. D29011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29011G Supplier TCI America Supplier No. D29011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6790-09-6
Molecular Formula C24H50O13
Molecular Weight (g/mol) 546.65
MDL Number MFCD06201001
InChI Key WRZXKWFJEFFURH-UHFFFAOYSA-N
PubChem CID 81248
ChEBI CHEBI:39524
IUPAC Name 2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO)O

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 325°C
Quantity 1 g
Formula Weight 546.65
Percent Purity ≥90.0% (GC)
Physical Form Wax
Chemical Name or Material Dodecaethylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.