Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecamethylcyclohexasiloxane 97.0+%, TCI America™
SDP

Catalog No. D204025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D204025ML 25 mL
D20401ML 1 mL
D20405ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D204025ML Supplier TCI America Supplier No. D204025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 540-97-6
Molecular Formula C12H36O6Si6
Molecular Weight (g/mol) 444.924
MDL Number MFCD00144215
InChI Key IUMSDRXLFWAGNT-UHFFFAOYSA-N
PubChem CID 10911
IUPAC Name 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)C

Specifications

Color Colorless
Boiling Point 245°C
Quantity 25 mL
Formula Weight 444.92
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecamethylcyclohexasiloxane
RTECSNumber GW2328550
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.