Learn More
Medchemexpress LLC Dodecamethylpentasiloxane | 141-63-9 | 96.97% | 384.84 | 10 ML
SDP
Catalog No. 5000366461
Encompass_Preferred
Dodecamethylpentasiloxane is a component of siloxanes and can be used as silicone oil. It exhibits insecticidal activity against bed bugs and appears as a colorless to light yellow liquid with a density of 0.875 g/cm³. This product is for research use only.
  • Component of siloxanes
  • Can be used as silicone oil
  • Exhibits insecticidal activity against bed bugs
  • Colorless to light yellow liquid
  • Density of 0.875 g/cm³
  • For research use only
Catalog No. 50-003-66461 Supplier Medchemexpress LLC Supplier No. HYW01103510ML
Dodecamethylpentasiloxane is a component of siloxanes and can be used as silicone oil. It exhibits insecticidal activity against bed bugs and appears as a colorless to light yellow liquid with a density of 0.875 g/cm³. This product is for research use only.
  • Component of siloxanes
  • Can be used as silicone oil
  • Exhibits insecticidal activity against bed bugs
  • Colorless to light yellow liquid
  • Density of 0.875 g/cm³
  • For research use only