Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecanal | 112-54-9 | 184.32 | 1 G
SDP

Catalog No. 5000448829
Encompass_Preferred

Dodecanal is a composition of essential oil that can be found in Coriandrum sativum L.

  • Molecular formula: C12H24O
  • Molecular weight: 184.32
  • Appearance: Liquid, colorless to light yellow
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-48829 Supplier Medchemexpress LLC Supplier No. HYW0043011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dodecanal is a composition of essential oil that can be found in Coriandrum sativum L.

  • Molecular formula: C12H24O
  • Molecular weight: 184.32
  • Appearance: Liquid, colorless to light yellow
  • Store at -20°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.