Learn More
Medchemexpress LLC Dodecane-1,12-diol | 5675-51-4 | MFCD00004755 | 99.2% | 202.34 g/mol | C12H26O2 | 1 ML
SDP
Catalog No. 5000302720
Encompass_Preferred
Dodecane-1,12-diol is a linear C12 diol with terminal hydroxyl groups used as a reagent and building block in polymer chemistry, enzymatic synthesis, and materials research. It is supplied as a solid or as a ready-to-use solution for convenient formulation and screening workflows.
  • High purity: 99.2% suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Good solubility in DMSO (≈50 mg/mL) for easy dissolution and handling.
  • Stable when stored under recommended conditions for extended shelf life.
  • White to off-white solid with molecular formula C12H26O2 and molecular weight 202.34 g/mol.
Catalog No. 50-003-02720 Supplier Medchemexpress LLC Supplier No. HYW01477910MM/1ML
Dodecane-1,12-diol is a linear C12 diol with terminal hydroxyl groups used as a reagent and building block in polymer chemistry, enzymatic synthesis, and materials research. It is supplied as a solid or as a ready-to-use solution for convenient formulation and screening workflows.
  • High purity: 99.2% suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Good solubility in DMSO (≈50 mg/mL) for easy dissolution and handling.
  • Stable when stored under recommended conditions for extended shelf life.
  • White to off-white solid with molecular formula C12H26O2 and molecular weight 202.34 g/mol.