Learn More
Medchemexpress LLC Dodecane-1,12-diol | 5675-51-4 | MFCD00004755 | 99.2% | 202.34 g·mol⁻¹ | C12H26O2 | 25 G
SDP
Catalog No. 5000303067
Encompass_Preferred
Dodecane-1,12-diol is a linear C12 primary diol supplied as a high-purity solid for use as an organic reagent and building block in polymer and biochemical synthesis. It is commonly used where a long-chain diol is required for polyester production, formulation chemistry, and organic transformations.
  • High purity (99.22%).
  • Linear C12 diol useful as a monomer and building block.
  • Solid form suitable for storage and handling.
  • Applicable to polyester synthesis, surfactant, and lubricant formulation.
  • Available in multiple packaging sizes (25 g-500 g and bulk inquiry).
Catalog No. 50-003-03067 Supplier Medchemexpress LLC Supplier No. 25GHYW014779
Dodecane-1,12-diol is a linear C12 primary diol supplied as a high-purity solid for use as an organic reagent and building block in polymer and biochemical synthesis. It is commonly used where a long-chain diol is required for polyester production, formulation chemistry, and organic transformations.
  • High purity (99.22%).
  • Linear C12 diol useful as a monomer and building block.
  • Solid form suitable for storage and handling.
  • Applicable to polyester synthesis, surfactant, and lubricant formulation.
  • Available in multiple packaging sizes (25 g-500 g and bulk inquiry).