Learn More
Medchemexpress LLC Dodecane-1,12-diol | 5675-51-4 | MFCD00004755 | 99.2% | 202.34 g/mol | C12H26O2 | 50 G
SDP
Catalog No. 5000302721
Encompass_Preferred
Dodecane-1,12-diol (dodecamethylene glycol) is a linear C12 diol used as a chemical building block and biochemical reagent. It is commonly employed in the synthesis of polyesters and other polymers, and in enzymatic reactions for producing bio-based materials. The compound is supplied as a solid with high purity suitable for research and laboratory use.
  • Linear C12 diol useful as a monomer in polymer synthesis.
  • Supports enzymatic synthesis of bio-based polyesters.
  • High purity suitable for research applications.
  • Solid form with molecular formula C12H26O2 and MW 202.34 g/mol.
  • Available in multiple laboratory package sizes, including 50 g.
Catalog No. 50-003-02721 Supplier Medchemexpress LLC Supplier No. HYW01477950G
Dodecane-1,12-diol (dodecamethylene glycol) is a linear C12 diol used as a chemical building block and biochemical reagent. It is commonly employed in the synthesis of polyesters and other polymers, and in enzymatic reactions for producing bio-based materials. The compound is supplied as a solid with high purity suitable for research and laboratory use.
  • Linear C12 diol useful as a monomer in polymer synthesis.
  • Supports enzymatic synthesis of bio-based polyesters.
  • High purity suitable for research applications.
  • Solid form with molecular formula C12H26O2 and MW 202.34 g/mol.
  • Available in multiple laboratory package sizes, including 50 g.