Learn More
Medchemexpress LLC Dodecane-1,12-diol | 5675-51-4 | MFCD00004755 | 99.2% | 202.34 g/mol | C12H26O2 | 500 G
SDP
Catalog No. 5000302121
Encompass_Preferred
Dodecane-1,12-diol is a linear aliphatic diol (C12H26O2, CAS 5675-51-4) used as a biochemical reagent and synthetic building block in organic chemistry. Supplied as a solid with high reported purity, it is intended for laboratory research and not for human use.
  • High purity (99.22%) suitable for research applications.
  • Linear diol useful as a building block for polyesters and organic synthesis.
  • Solid form with molecular weight ≈202.34 g/mol and formula C12H26O2.
  • Available in multiple pack sizes including 500 g for scale-up experiments.
  • Provided with safety disclaimer for research use only and not for patient use.
Catalog No. 50-003-02121 Supplier Medchemexpress LLC Supplier No. HYW014779500G
Dodecane-1,12-diol is a linear aliphatic diol (C12H26O2, CAS 5675-51-4) used as a biochemical reagent and synthetic building block in organic chemistry. Supplied as a solid with high reported purity, it is intended for laboratory research and not for human use.
  • High purity (99.22%) suitable for research applications.
  • Linear diol useful as a building block for polyesters and organic synthesis.
  • Solid form with molecular weight ≈202.34 g/mol and formula C12H26O2.
  • Available in multiple pack sizes including 500 g for scale-up experiments.
  • Provided with safety disclaimer for research use only and not for patient use.