Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecane-1,12-diol-d24 | 1219802-88-6 | 99.0% | 226.48 | C12H2D24O2 | 1 MG
SDP

Catalog No. 5000302122
Encompass_Preferred

Dodecane-1,12-diol-d24 is the fully deuterated isotopologue of 1,12-dodecanediol supplied as a high-purity, stable-isotope labeled standard for analytical and synthetic research. It is intended for use as an internal standard, tracer, or labeled building block in mass spectrometry and isotopic studies, provided as a powder with defined storage conditions to preserve isotopic enrichment.

  • Fully deuterated (d24) isotopologue with high isotopic enrichment.
  • 99.0% isotopic purity as specified by the manufacturer.
  • Molecular formula C12H2D24O2; molecular weight 226.48.
  • Suitable for mass spectrometry standards and metabolic tracing.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Available in small analytical quantities for research use.

Catalog No. 50-003-02122 Supplier Medchemexpress LLC Supplier No. HYW014779S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dodecane-1,12-diol-d24 is the fully deuterated isotopologue of 1,12-dodecanediol supplied as a high-purity, stable-isotope labeled standard for analytical and synthetic research. It is intended for use as an internal standard, tracer, or labeled building block in mass spectrometry and isotopic studies, provided as a powder with defined storage conditions to preserve isotopic enrichment.

  • Fully deuterated (d24) isotopologue with high isotopic enrichment.
  • 99.0% isotopic purity as specified by the manufacturer.
  • Molecular formula C12H2D24O2; molecular weight 226.48.
  • Suitable for mass spectrometry standards and metabolic tracing.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Available in small analytical quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.