Learn More
Medchemexpress LLC Dodecane-1,12-diol-d24 | 1219802-88-6 | MFCD08705053 | 99.0% | 226.48 g/mol | C12H2D24O2 | 5 MG
SDP
Catalog No. 5000303275
Encompass_Preferred
Dodecane-1,12-diol-d24 is the deuterium-labeled analogue of dodecane-1,12-diol provided as a stable-isotope reagent for research applications. It is intended for use as an internal standard, tracer, or isotopically labeled building block in analytical chemistry and synthetic workflows.
  • High isotopic enrichment suitable for mass spectrometry and NMR studies.
  • High chemical purity (99%) for reproducible analytical results.
  • White to off-white solid, easy to handle and weigh.
  • Soluble in common organic solvents for sample preparation and synthesis.
  • Available in small, accurately quantified quantities for method development.
Catalog No. 50-003-03275 Supplier Medchemexpress LLC Supplier No. HYW014779S5MG
Dodecane-1,12-diol-d24 is the deuterium-labeled analogue of dodecane-1,12-diol provided as a stable-isotope reagent for research applications. It is intended for use as an internal standard, tracer, or isotopically labeled building block in analytical chemistry and synthetic workflows.
  • High isotopic enrichment suitable for mass spectrometry and NMR studies.
  • High chemical purity (99%) for reproducible analytical results.
  • White to off-white solid, easy to handle and weigh.
  • Soluble in common organic solvents for sample preparation and synthesis.
  • Available in small, accurately quantified quantities for method development.