Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018375 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8375 25 mL
11-101-9799 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8375 Supplier MilliporeSigma Supelco Supplier No. 4401025ML
Only null left
Add to Cart
Add to Cart

Dodecane is a clear, colorless liquid, having a mild aliphatic hydrocarbon odor, which belongs to the family of acyclic alkanes and contains compounds like 3-methylundecane and 5-methylundecane.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 401°F
CAS 112-40-3
Vapor Pressure 1 mmHg (47.8°C)
Vapor Density 5.96 (vs. air)
Density 0.75 g/mL (at 25°C)
Flash Point 74°C
Physical Form Neat
Linear Formula CH3(CH2)10 CH3
MDL Number MFCD00008969
Melting Point -9.6°C
Boiling Point 215°C to 217°C
Type Analytical Standard
Quantity 25 mL
Formula Weight 170.33
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.