Learn More
Medchemexpress LLC Dodecanedioic acid | 693-23-2 | ≥98.0% | 230.30 | 100 G
SDP
Catalog No. 5000453009
Encompass_Preferred
Dodecanedioic acid is an orally active straight-chain dicarboxylic acid that serves as an alternative energy substrate between lipids and carbohydrates. It is rapidly oxidized during exercise, which helps reduce muscle fatigue without stimulating insulin secretion. Additionally, Dodecanedioic acid has been observed to lower blood glucose levels in individuals with non-insulin-dependent diabetes mellitus.
  • Orally active straight-chain dicarboxylic acid.
  • Alternative energy substrate between lipids and carbohydrates.
  • Rapidly oxidized during exercise, reducing muscle fatigue.
  • Does not stimulate insulin secretion.
  • Exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus.
Catalog No. 50-004-53009 Supplier Medchemexpress LLC Supplier No. HYW012241100G
Add to Cart
Add to Cart
Dodecanedioic acid is an orally active straight-chain dicarboxylic acid that serves as an alternative energy substrate between lipids and carbohydrates. It is rapidly oxidized during exercise, which helps reduce muscle fatigue without stimulating insulin secretion. Additionally, Dodecanedioic acid has been observed to lower blood glucose levels in individuals with non-insulin-dependent diabetes mellitus.
  • Orally active straight-chain dicarboxylic acid.
  • Alternative energy substrate between lipids and carbohydrates.
  • Rapidly oxidized during exercise, reducing muscle fatigue.
  • Does not stimulate insulin secretion.
  • Exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus.