Learn More
Medchemexpress LLC Dodecanedioic acid-d4 | 97543-02-7 | 234.33 | 10 MG
SDP
Catalog No. 5000452620
Encompass_Preferred
Dodecanedioic acid-d4 is the deuterium labeled version of Dodecanedioic acid. Dodecanedioic acid is an orally active straight-chain dicarboxylic acid that acts as an alternative energy substrate. It can be rapidly oxidized during exercise, reducing muscle fatigue without stimulating insulin secretion, and also exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus.
  • Deuterium labeled compound for quantitative tracing during drug development.
  • Reduces muscle fatigue during exercise.
  • Exhibits blood glucose-lowering activity.
Catalog No. 50-004-52620 Supplier Medchemexpress LLC Supplier No. HYW012241S110MG
Dodecanedioic acid-d4 is the deuterium labeled version of Dodecanedioic acid. Dodecanedioic acid is an orally active straight-chain dicarboxylic acid that acts as an alternative energy substrate. It can be rapidly oxidized during exercise, reducing muscle fatigue without stimulating insulin secretion, and also exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus.
  • Deuterium labeled compound for quantitative tracing during drug development.
  • Reduces muscle fatigue during exercise.
  • Exhibits blood glucose-lowering activity.