Learn More
Medchemexpress LLC Dodecanoic-1-13C acid | 93639-08-8 | 99.8% | 201.31 g·mol⁻1 | C11(13C)H24O2 | 25mg
SDP
Catalog No. 5000269693
Encompass_Preferred
Lauric acid-13C is the 13C labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-69693 Supplier Medchemexpress LLC Supplier No. HYY0366S25MG
Lauric acid-13C is the 13C labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively