Learn More
Medchemexpress LLC Dodecanoic-12,12,12-d3 acid | 79050-22-9 | MFCD00190437 | 98.9% | 203.34 g/mol | C12H21D3O2 | 5mg
SDP
Catalog No. 5000268281
Encompass_Preferred
Lauric acid-d33 is the deuterium labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-68281 Supplier Medchemexpress LLC Supplier No. HYY0366S35MG
Lauric acid-d33 is the deuterium labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively