Learn More
Medchemexpress LLC Dodecanoic-2,2-d2 acid | 64118-39-4 | MFCD00053326 | 99.8% | 202.33 | C12H22D2O2 | 100mg
SDP
Catalog No. 5000269694
Encompass_Preferred
Lauric acid-d2 is the deuterium labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-69694 Supplier Medchemexpress LLC Supplier No. HYY0366S2100MG
Lauric acid-d2 is the deuterium labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively