Learn More
Medchemexpress LLC Dodecanoic acid | 143-07-7 | MFCD00002736 | 99.7% | 200.32 g/mol | C12H24O2 | 250mg
SDP
Catalog No. 5000268226
Encompass_Preferred
Lauric acid (Standard) is the analytical standard of Lauric acid This product is intended for research and analytical applications Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-68226 Supplier Medchemexpress LLC Supplier No. HYY0366R250MG
Lauric acid (Standard) is the analytical standard of Lauric acid This product is intended for research and analytical applications Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively