Learn More
Medchemexpress LLC Dodecanoic acid | 143-07-7 | MFCD00002736 | 99.9% | 200.32 | C12H24O2 | 50mg
SDP
Catalog No. 5000268217
Encompass_Preferred
Lauric acid (Standard) is the analytical standard of Lauric acid This product is intended for research and analytical applications Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-68217 Supplier Medchemexpress LLC Supplier No. HYY0366R50MG
Lauric acid (Standard) is the analytical standard of Lauric acid This product is intended for research and analytical applications Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively