Learn More
Medchemexpress LLC Dodecanoic acid (lauric acid) | 143-07-7 | MFCD00002736 | 99.9% | 200.32 g/mol | C12H24O2 | 100g
SDP
Catalog No. 5000269955
Encompass_Preferred
Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively[1]
Catalog No. 50-002-69955 Supplier Medchemexpress LLC Supplier No. HYY0366100G
Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively[1]