Learn More
Apexbio Technology LLC Dodecanoyl-carnitine-d3-HCl
SDP
Catalog No. 5000570235
Encompass_Preferred
Dodecanoyl carnitine-d3 HCl (CAS No 2687960-76-3) is a deuterated analog of Dodecanoyl carnitine used in biomedical research particularly in metabolism and lipid biochemistry studies As a derivative of carnitine it is instrumental in investigating fatty acid metabolism and energy production pathways The compound is often utilized to probe mitochondrial function and the activity of carnitine-related transport mechanisms It exhibits activity in the nanomolar to low micromolar range in vitro Typical applications include use in cell culture models animal studies and drug screening assays for metabolic disorders Its experimental concentrations are typically dependent on the specific research design allowing flexibility for various experimental setups
Catalog No. 50-005-70235 Supplier Apexbio Technology LLC Supplier No. SL102710
Dodecanoyl carnitine-d3 HCl (CAS No 2687960-76-3) is a deuterated analog of Dodecanoyl carnitine used in biomedical research particularly in metabolism and lipid biochemistry studies As a derivative of carnitine it is instrumental in investigating fatty acid metabolism and energy production pathways The compound is often utilized to probe mitochondrial function and the activity of carnitine-related transport mechanisms It exhibits activity in the nanomolar to low micromolar range in vitro Typical applications include use in cell culture models animal studies and drug screening assays for metabolic disorders Its experimental concentrations are typically dependent on the specific research design allowing flexibility for various experimental setups