Learn More
TARGETMOL CHEMICALS INC Dodecanoylcarnitine 25MG
SDP
Catalog No. 502878602
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dodecanoylcarnitine ((-)-Lauroylcarnitine) has been associated with disturbances in fatty acid oxidation, and the use of Dodecanoylcarnitine resulted in an increase in the permeability of salmon calcitonin. Purity 99.72%
Catalog No. 50-287-8602 Supplier TARGETMOL CHEMICALS INC Supplier No. T1515725MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dodecanoylcarnitine ((-)-Lauroylcarnitine) has been associated with disturbances in fatty acid oxidation, and the use of Dodecanoylcarnitine resulted in an increase in the permeability of salmon calcitonin. Purity 99.72%