Learn More
Crescent Chemical Co Inc DODECENYLSUCCINIC ACID
SDP
Supplier:  Crescent Chemical Co Inc SE20755.02
Encompass_Preferred
Dodecenylsuccinic acid anhydride pract. Hardener for epoxy resins. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 20755.02 CAS 25377-73-5
Catalog No. 50-247-536