Learn More
Cayman Chemical Dodecanoyl D-Sucrose, Purity: ≥95%, CAS: 25339-99-5, 500 mg
SDP
Catalog No. 502989787
Encompass_Preferred
A nonionic surfactant; active against B. cereus, B. subtilis, S. aureus, E. coli, and S. typhimurium in a disc diffusion assay at 255 mM; immunization of mice with dodecanoyl D-sucrose-coated ovalbumin increases serum IgG levels by 5-fold compared to uncoated ovalbumin; dodecanoyl D-sucrose hydrogel (15% v/v) containing estradiol increases serum estradiol levels in rabbits compared to an ethanol-containing hydrogel; has been used to prepare stable foams in aqueous solutions, as well as oil-in-water emulsions
Catalog No. 50-298-9787 Supplier Cayman Chemical Supplier No. 29982500 MG
A nonionic surfactant; active against B. cereus, B. subtilis, S. aureus, E. coli, and S. typhimurium in a disc diffusion assay at 255 mM; immunization of mice with dodecanoyl D-sucrose-coated ovalbumin increases serum IgG levels by 5-fold compared to uncoated ovalbumin; dodecanoyl D-sucrose hydrogel (15% v/v) containing estradiol increases serum estradiol levels in rabbits compared to an ethanol-containing hydrogel; has been used to prepare stable foams in aqueous solutions, as well as oil-in-water emulsions