Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Alcohol, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610309 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-309 3 kg Amber Glass Bottle
18-610-310 100 g Amber Glass Bottle
18-610-312 500 g Amber Glass Bottle
18-610-311 20 L Amber Glass Bottle
18-610-313 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-309 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D10673KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

CAS Max % 1
Boiling Point 254°C to 261°C
Packaging Amber Glass Bottle
Quantity 3 kg
Percent Purity 0.98
Grade ACS
Chemical Name or Material Dodecyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.