Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Dodecyl Alcohol
SCI_ED

Catalog No. S25686A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25686A 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25686A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 112-53-8
Color White
Density 0.835g/mL
Packaging Solid
Molecular Formula C12H26O
Linear Formula CH3(CH2)11OH
MDL Number MFCD00004753
Quantity 100 g
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCO
IUPAC Name dodecan-1-ol
Molecular Weight (g/mol) 186.34
PubChem CID 8193
ChEBI CHEBI:28878
Formula Weight 186.34
Chemical Name or Material Dodecyl Alcohol
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.