Learn More
Medchemexpress LLC Dodecyl azide | 13733-78-3 | >98.0% | 211.35 g·mol⁻¹ | C12H25N3 | 1 G
SDP
Catalog No. 5000284278
Encompass_Preferred
Dodecyl azide (1-azidododecane) is an organic azide reagent bearing a 12-carbon alkyl chain. The terminal azide (N3) participates in bioorthogonal click chemistry with alkynes, BCN, and DBCO. Supplied for research use only; molecular formula C12H25N3, molecular weight 211.35 g·mol⁻¹.
  • Terminal azide group for azide-alkyne cycloaddition reactions.
  • Hydrophobic 12-carbon chain suitable for labeling and surface modification.
  • Available in common laboratory pack sizes, including 1 g.
  • Store solid at -20°C for long-term stability; store solutions at -80°C for extended storage.
  • Provided for research use only; not for human use.
Catalog No. 50-002-84278 Supplier Medchemexpress LLC Supplier No. HYW5905431G
Dodecyl azide (1-azidododecane) is an organic azide reagent bearing a 12-carbon alkyl chain. The terminal azide (N3) participates in bioorthogonal click chemistry with alkynes, BCN, and DBCO. Supplied for research use only; molecular formula C12H25N3, molecular weight 211.35 g·mol⁻¹.
  • Terminal azide group for azide-alkyne cycloaddition reactions.
  • Hydrophobic 12-carbon chain suitable for labeling and surface modification.
  • Available in common laboratory pack sizes, including 1 g.
  • Store solid at -20°C for long-term stability; store solutions at -80°C for extended storage.
  • Provided for research use only; not for human use.