Learn More
Medchemexpress LLC Dodecyl azide | 13733-78-3 | MFCD00223067 | ≥98.0% | 211.35 g·mol⁻¹ | C12H25N3 | 500 MG
SDP
Catalog No. 5000284587
Encompass_Preferred
Dodecyl azide (1-azidododecane) is an aliphatic organic azide bearing a 12-carbon hydrophobic tail and a reactive terminal azide (N3) group. It is used as a lipophilic clickable handle for copper-catalyzed and strain-promoted azide-alkyne cycloadditions (for example with alkynes, BCN, or DBCO). Molecular formula C12H25N3; molecular weight 211.35 g·mol⁻¹. Store under cold conditions per supplier guidance.
  • Terminal azide functional group for click chemistry.
  • 12-carbon hydrophobic tail for incorporation into lipophilic environments.
  • Suitable for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High purity as indicated on batch certificate of analysis.
  • Supplied in laboratory pack sizes for synthesis and labeling workflows.
Catalog No. 50-002-84587 Supplier Medchemexpress LLC Supplier No. HYW590543500MG
Dodecyl azide (1-azidododecane) is an aliphatic organic azide bearing a 12-carbon hydrophobic tail and a reactive terminal azide (N3) group. It is used as a lipophilic clickable handle for copper-catalyzed and strain-promoted azide-alkyne cycloadditions (for example with alkynes, BCN, or DBCO). Molecular formula C12H25N3; molecular weight 211.35 g·mol⁻¹. Store under cold conditions per supplier guidance.
  • Terminal azide functional group for click chemistry.
  • 12-carbon hydrophobic tail for incorporation into lipophilic environments.
  • Suitable for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High purity as indicated on batch certificate of analysis.
  • Supplied in laboratory pack sizes for synthesis and labeling workflows.