Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecyl dihydrogen phosphate | 2627-35-2 | MFCD00013946 | >95.0% | 266.31 g/mol | C12H27O4P | 50MG
SDP

Catalog No. 5000238007
Encompass_Preferred

Monolauryl phosphate (dodecyl dihydrogen phosphate) is a biologically active organophosphate reagent used in biochemical research and assay applications. It is the monododecyl ester of phosphoric acid, typically supplied as a solid with supporting documentation for purity and safe handling. Small pack sizes are commonly available for laboratory use, and suppliers provide datasheets and safety data sheets to support experimental use and storage.

  • CAS number 2627-35-2.
  • Chemical formula C12H27O4P.
  • Molecular weight 266.31 g/mol.
  • High purity reported by suppliers (e.g., >95%).
  • Available in small laboratory pack sizes such as 50 mg, 100 mg, and 250 mg.
  • Product documentation includes data sheet and SDS for handling and safety.

Catalog No. 50-002-38007 Supplier Medchemexpress LLC Supplier No. HY12580250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monolauryl phosphate (dodecyl dihydrogen phosphate) is a biologically active organophosphate reagent used in biochemical research and assay applications. It is the monododecyl ester of phosphoric acid, typically supplied as a solid with supporting documentation for purity and safe handling. Small pack sizes are commonly available for laboratory use, and suppliers provide datasheets and safety data sheets to support experimental use and storage.

  • CAS number 2627-35-2.
  • Chemical formula C12H27O4P.
  • Molecular weight 266.31 g/mol.
  • High purity reported by suppliers (e.g., >95%).
  • Available in small laboratory pack sizes such as 50 mg, 100 mg, and 250 mg.
  • Product documentation includes data sheet and SDS for handling and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.