Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecyl gallate | 1166-52-5 | MFCD00002195 | 99.8% | 338.44 g/mol | C19H30O5 | 100MG
SDP

Catalog No. 5000236027
Encompass_Preferred

Dodecyl gallate (CAS 1166-52-5) is the analytical reference standard of lauryl gallate supplied for research and analytical use. The material is provided with high purity and characterized for formula and molecular weight, making it suitable for antioxidant studies and analytical assays.

  • Analytical reference standard for dodecyl (lauryl) gallate.
  • High purity, 99.8%.
  • Chemical formula C19H30O5.
  • Molecular weight 338.44 g/mol.
  • Available in small pack sizes, including 100 mg.
  • Intended for research and analytical applications.

Catalog No. 50-002-36027 Supplier Medchemexpress LLC Supplier No. HY124082R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dodecyl gallate (CAS 1166-52-5) is the analytical reference standard of lauryl gallate supplied for research and analytical use. The material is provided with high purity and characterized for formula and molecular weight, making it suitable for antioxidant studies and analytical assays.

  • Analytical reference standard for dodecyl (lauryl) gallate.
  • High purity, 99.8%.
  • Chemical formula C19H30O5.
  • Molecular weight 338.44 g/mol.
  • Available in small pack sizes, including 100 mg.
  • Intended for research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.