Learn More
Medchemexpress LLC Dodecyl gallate | 1166-52-5 | MFCD00002195 | 100.0% | 338.44 g/mol | C19H30O5 | 25G
SDP
Catalog No. 5000236956
Encompass_Preferred
Dodecyl gallate (lauryl gallate) is an alkyl gallate antioxidant used as a preservative and research reagent in food, pharmaceutical, and cosmetic applications. It is a white to off-white solid with reported antibacterial activity against Gram-positive bacteria and reported antiviral activity against African swine fever virus; commonly used in formulation, stability, and antimicrobial studies.
  • Antioxidant properties useful for formulation and stability testing.
  • Reported antibacterial activity against Gram-positive bacteria.
  • Reported antiviral activity relevant to veterinary research.
  • High reported purity for reproducible experimental results.
  • Available in laboratory-scale package sizes for research use.
Catalog No. 50-002-36956 Supplier Medchemexpress LLC Supplier No. HY12408225G
Dodecyl gallate (lauryl gallate) is an alkyl gallate antioxidant used as a preservative and research reagent in food, pharmaceutical, and cosmetic applications. It is a white to off-white solid with reported antibacterial activity against Gram-positive bacteria and reported antiviral activity against African swine fever virus; commonly used in formulation, stability, and antimicrobial studies.
  • Antioxidant properties useful for formulation and stability testing.
  • Reported antibacterial activity against Gram-positive bacteria.
  • Reported antiviral activity relevant to veterinary research.
  • High reported purity for reproducible experimental results.
  • Available in laboratory-scale package sizes for research use.