Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium dodecyl sulfate BioXtra, >=99.0% (GC) | 151-21-3 | MFCD00036175 | 250G
Catalog No. NC1141007
Encompass
Sodium dodecyl sulfate BioXtra, >=99.0% (GC) | Purity: >=99.0% (GC) | Mol Wt: 288.38 | 151-21-3 | MFCD00036175 | 250G
Catalog No. NC1141007 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L6026250G
Sodium dodecyl sulfate BioXtra, >=99.0% (GC) | Purity: >=99.0% (GC) | Mol Wt: 288.38 | 151-21-3 | MFCD00036175 | 250G