Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylamine hydrochloride, 99%

Catalog No. AC409250100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409250100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC409250100 Supplier Thermo Scientific Chemicals Supplier No. 409250100
  • $69.25
    $61.65 / Each of 1

    Save $7.60

    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-73-7
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00042017
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
PubChem CID 458426
IUPAC Name dodecan-1-amine;hydrochloride
SMILES [Cl-].CCCCCCCCCCCC[NH3+]

Specifications

Melting Point 186°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3(CH2)11NH2·HCl
Quantity 10 g
Beilstein 04, IV, 794
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform,benzene,,toluene,diethyl ether
Formula Weight 221.81
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dodecylamine hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-205-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.