Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dodecylbenzenesulfonic acid solution

Catalog No. 522953500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-295-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-295-3500ML Supplier Sigma Aldrich Supplier No. 522953500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dodecylbenzenesulfonic acid solution (DBSA) is a non-volatile and non-hygroscopic acid catalyst majorly used in the trans-esterification reaction.Dopant for conducting polymers strong acid catalyst for amino cross-linked coatings

Specifications

CAS 27176-87-0
Density 0.992 g/mL (at 25°C)
Boiling Point 82°C
Molecular Formula C18H30O3S
Linear Formula C12H25C6H4SO3H
Refractive Index n20/D 1.479
MDL Number MFCD00066677
Quantity 500 mL
Synonym Dodecylbenzenesulfonic acid
Molecular Weight (g/mol) 326.49
Biological Activity Central nervous system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.