Learn More
Enzo Life Sciences Dodecylphosphate (5mg). CAS: 2627-35-2
SDP
Supplier:  Enzo Life Sciences BMLLP1050005
Encompass_Preferred
Lysophosphatidic acid (LPA) receptor ligand. A specific agonist at LPA2 (EC50=700 nM) and antagonist at LPA Alternative name: Dihydrogen monolauryl phosphate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (15 mg/ml warm) or ethanol (2 mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-1158