Learn More
Cayman Chemical Dodecylphosphocholine, Purity: ≥95%, CAS: 29557-51-5, 250 mg
SDP
Catalog No. 502934266
Encompass_Preferred
A detergent; commonly used for membrane protein structure determination using NMR; has been used to generate membrane mimetic micelles and to characterize pore formation by the influenza M2 transmembrane domain within a synthetic membrane; micelles induce amyloid fibril formation by PrP (110-136); CMC = 1.5 mM
Catalog No. 50-293-4266 Supplier Cayman Chemical Supplier No. 25629250 MG
A detergent; commonly used for membrane protein structure determination using NMR; has been used to generate membrane mimetic micelles and to characterize pore formation by the influenza M2 transmembrane domain within a synthetic membrane; micelles induce amyloid fibril formation by PrP (110-136); CMC = 1.5 mM