Learn More
Cayman Chemical Dodecylphosphocholine-d38, Purity: ≥99% deuterated forms (d1-d38), CAS: 130890-78-7, 250 mg
SDP
Catalog No. 502935657
Encompass_Preferred
An internal standard for the quantification of dodecylphosphocholine by GC- or LC-MS
Catalog No. 50-293-5657 Supplier Cayman Chemical Supplier No. 25996250 MG
An internal standard for the quantification of dodecylphosphocholine by GC- or LC-MS