Learn More
Crescent Chemical Co Inc DODECYLSULFATE-NA-SALT
SDP
Supplier:  Crescent Chemical Co Inc SE20765.01
Encompass_Preferred
Dodecylsulfate-Na-salt in pellets research grade Ultrapure SDS pressed in small pellets thus avoiding the irritant dust of the powder form. Suitable for SDS PAGE and biochemistry. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 20765.01 CAS No
Catalog No. 50-247-544