Learn More
Dodecyltrimethylammonium bromide, For ion pair chromatography, ≥98.5% (AT), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Melting Point | 246°C |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | CH3(CH2)11 N(CH3)3 Br |
| Formula Weight | 308.34 |
| Percent Purity | ≥98.5% (AT) |
| Grade | Ion Pair Chromatography |
| Chemical Name or Material | Dodecyltrimethylammonium Bromide |
Safety and Handling
| RTECSNumber | BQ3195000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.