Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecyltrimethylammonium-d3 chloride | 1219337-20-8 | MFCD28899604 | 266.91 g·mol⁻¹ | C15H31D3ClN | 10 MG
SDP

Catalog No. 5000284663
Encompass_Preferred

DTAC-d3 (Dodecyltrimethylammonium-d3 chloride) is a deuterium-labeled analogue of DTAC supplied as a biochemical reagent for life-science research. It is commonly used as an isotope-labeled tracer or internal standard in analytical workflows, including mass spectrometry and lipidomics, and is supplied as a solid for laboratory use.

  • Deuterium-labeled internal standard for analytical assays.
  • Quaternary ammonium salt suitable for tracer and isotope studies.
  • Molecular formula: C15H31D3ClN; molecular weight: 266.91 g·mol⁻¹.
  • CAS number: 1219337-20-8.
  • Certificate of analysis provides lot-specific purity information.

Catalog No. 50-002-84663 Supplier Medchemexpress LLC Supplier No. HYW70077010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

DTAC-d3 (Dodecyltrimethylammonium-d3 chloride) is a deuterium-labeled analogue of DTAC supplied as a biochemical reagent for life-science research. It is commonly used as an isotope-labeled tracer or internal standard in analytical workflows, including mass spectrometry and lipidomics, and is supplied as a solid for laboratory use.

  • Deuterium-labeled internal standard for analytical assays.
  • Quaternary ammonium salt suitable for tracer and isotope studies.
  • Molecular formula: C15H31D3ClN; molecular weight: 266.91 g·mol⁻¹.
  • CAS number: 1219337-20-8.
  • Certificate of analysis provides lot-specific purity information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.