Learn More
Cayman Chemical Dodecyltrimethylammonium (bromide), Purity: ≥98%, CAS: 1119-94-4, 5 g
SDP
Catalog No. 502915356
Encompass_Preferred
A quaternary ammonium-containing cationic surfactant; CMC = 15.7 and 8.1 mM in water and PBS, pH 7.4, respectively; binds to DNA; active against L. monocytogenes (MIC = 120 mg/L)
Catalog No. 50-291-5356 Supplier Cayman Chemical Supplier No. 370095 G
A quaternary ammonium-containing cationic surfactant; CMC = 15.7 and 8.1 mM in water and PBS, pH 7.4, respectively; binds to DNA; active against L. monocytogenes (MIC = 120 mg/L)